missing translation for 'onlineSavingsMsg'
Learn More

FGFR5/FGFRL1 Antibody (004), DyLight 405, Novus Biologicals™

Product Code. 18327630 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327630 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327630 Supplier Novus Biologicals Supplier No. NBP290466V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FGFR5/FGFRL1 Monoclonal specifically detects FGFR5/FGFRL1 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFR5/FGFRL1
Applications ELISA
Classification Monoclonal
Clone 4
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias FGF homologous factor receptor, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse FGFR5/FGFRL1 (NP_473412.1; Met 1-Pro 374).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Gene ID (Entrez) 53834
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.