missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGFR4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529596
This item is not returnable.
View return policy
Description
Recommended positive controls: Huh-7, HepG2, HepG2 membrane extract. Predicted reactivity: Mouse (86%), Rat (87%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FGFR4 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy. FGFR4 is required for normal down-regulation of the expression of CYP7A1, the rate-limiting enzyme in bile acid synthesis, in response to FGF19. FGFR4 plays a role in postnatal lung development and may be involved in the development of skeletal muscle cell lineages.
Specifications
| FGFR4 | |
| Polyclonal | |
| Unconjugated | |
| Fgfr4 | |
| CD334; CTLA-2-beta protein; FGF Receptor; FGF Receptor 4; FGFR; Fgfr4; FGFR-4; Fibroblast growth factor receptor 4; fibroblast growth factor receptor 4 16 minus form; hydroxyaryl-protein kinase; JTK2; MGC20292; Mpk-11; protein-tyrosine kinase; protein-tyrosine kinase receptor MPK-11; TKF; tyrosine kinase related to fibroblast growth factor receptor; tyrosylprotein kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2264 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.95 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P22455 | |
| Fgfr4 | |
| Recombinant protein encompassing a sequence within the center region of human FGFR4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction