missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FGFR4/CEN5q FISH Probe

Product Code. p-7190307
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
This item is not returnable. View return policy

Product Code. 16083065

Brand: Abnova™ FG0051.200uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

DAPI Counterstain (1500ng/mL) 250μL

Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.

FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.

Advantages

  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity

Specifications

Content And Storage Store at 4°C in the dark.
Label or Dye FITC, Texas Red
For Use With (Application) Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
Includes DAPI Counterstain (1500 ng/mL ) 250 uL
Location 5q35.1, 5q11.2
Probe FGFR4, CEN5q
Product Type FGFR4/CEN5q FISH Probe
Quantity 200 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.