missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FGFR4 Antibody (U3-1784), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28842-100ug
This item is not returnable.
View return policy
Description
FGFR4 Monoclonal antibody specifically detects FGFR4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| FGFR4 | |
| Monoclonal | |
| Unconjugated | |
| P22455 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| U3-1784 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase | |
| FGFR4/ CD334 | |
| 100 μg | |
| Angiogenesis, Protein Kinase | |
| 2264 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction