missing translation for 'onlineSavingsMsg'
Learn More

FGFR4 Antibody (4FR6D3) - BSA Free, Novus Biologicals™

Product Code. 18394012 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394012 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394012 Supplier Novus Biologicals Supplier No. NBP309137

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGFR4 Monoclonal specifically detects FGFR4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFR4
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 4FR6D3
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 2-6 ug/ml, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias CD334, CD334 antigen, EC 2.7.10, EC 2.7.10.1, FGFR-4, fibroblast growth factor receptor 4, JTK2hydroxyaryl-protein kinase, MGC20292, protein-tyrosine kinase, TKF, tyrosine kinase related to fibroblast growth factor receptor, tyrosylprotein kinase
Host Species Mouse
Immunogen NIH 3T3 cells transfected with full length human FGFR4
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2264
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.