missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGFR3 Recombinant Rabbit Monoclonal Antibody (HL1595)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556707
This item is not returnable.
View return policy
Description
FGFR3 Recombinant Monoclonal Antibody for Western Blot
FGFR3 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein would consist of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. This particular family member binds acidic and basic fibroblast growth hormone and plays a role in bone development and maintenance. Mutations in this gene lead to craniosynostosis and multiple types of skeletal dysplasia. Three alternatively spliced transcript variants that encode different protein isoforms have been described.
Specifications
| FGFR3 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with no preservative | |
| P22607 | |
| FGFR3 | |
| Synthetic peptide encompassing a sequence within the Intracellular domain of human FGFR3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Canine, Feline | |
| Antibody | |
| IgG |
| Western Blot | |
| HL1595 | |
| Unconjugated | |
| FGFR3 | |
| ACH; CD333; CEK2; FGF Receptor; FGFR; FGFR3; FGFR-3; FGFR3deltaTM; Fibroblast growth factor receptor 3; fibroblast growth factor receptor 3 variant 4; Flg-2; FR3; HBGFR; heparin-binding growth factor receptor; HSFGFR3EX; hydroxyaryl-protein kinase; JTK4; Mfr3; sam3; tyrosine kinase JTK4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100141386, 2261, 488808 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction