missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR3 Rabbit anti-Mouse, Clone: 46, Novus Biologicals™
Rabbit Monoclonal Antibody
£296.00 - £420.00
Specifications
| Antigen | FGFR3 |
|---|---|
| Clone | 46 |
| Dilution | ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18249377
|
Novus Biologicals
NBP2-90393-50UL |
50 μg |
£296.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18223856
|
Novus Biologicals
NBP2-90393-100UL |
100 μg |
£420.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGFR3 Monoclonal specifically detects FGFR3 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
| FGFR3 | |
| ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| Recombinant Mouse FGFR3 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 46 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Protein Kinase | |
| 0.2 um filtered solution in PBS | |
| 2261 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title