Learn More
FGFR3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55358-1-AP-150UL
This item is not returnable.
View return policy
Description
The FGFR3 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human FGFR3. This antibody recognizes human, mouse, rat antigen. The FGFR3 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| FGFR3 | |
| 650 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Immunohistochemistry, ELISA | |
| ACH, CD333, CEK2, FGFR 3, FGFR3, HSFGFR3EX, JTK4 | |
| 150 μL | |
| 2261 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |