missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX FGFR3 Protein, Active

Product Code. 17380204
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17380204

Brand: Eurofins DiscoverX 14464

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, recombinant human FGFR residues 447-761, containing an N-terminal His6-tag, for use in Kinase Assays.

Fibroblast growth factor (FGF) superfamily consists of 23 members, all of which contain a conserved 120 amino acid (aa) core region that contains six identical, interspersed amino acids. Based on the pI factor, two main groups of FGF are named: acidic FGF (aFGF) and basic FGF (bFGF). The superfamily members act extracellularly through four tyrosine kinase FGF receptors (FGR1 through FGR4), with multiple specificities noted for almost all FGFs. Biological binding of aFGF to its receptors will result in phosphorylation of the receptor and non-receptor proteins and generate its function. FGR3 (FGFR-3, Fibroblast growth factor receptor 3, EC 2.7.1.112) is expressed mainly in bone, central nervous system and tumors. Inhibition of FGR3 may be useful in the treatment of dwarfism as well as some tumors.

GenBank M58051

Specifications

Formulation mg/mL in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 36.9 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in SF21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥60%
Protein FGFR3, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.