missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR3, p Tyr724 Antibody (SR2305), Novus Biologicals™
Rabbit Monoclonal Antibody
£282.00 - £367.00
Specifications
| Antigen | FGFR3 |
|---|---|
| Clone | SR2305 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694205
|
Novus Biologicals
NBP3-21595-100ul |
100 μg |
£367.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18604035
|
Novus Biologicals
NBP3-21595-50ul |
50 μg |
£282.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGFR3 Monoclonal antibody specifically detects FGFR3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
| FGFR3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| A synthesized peptide derived from human phospho-FGFR3 [Tyr724] (Uniprot #: P22607) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2305 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Protein Kinase | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 2261 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title