missing translation for 'onlineSavingsMsg'
Learn More

FGFR3 Antibody (136318) [DyLight 550], Novus Biologicals™

Product Code. 30735660 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30735660 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735660 Supplier Novus Biologicals Supplier No. FAB7661L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGFR3 Monoclonal antibody specifically detects FGFR3 in Human samples. It is validated for Western Blot,Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFR3
Applications Western Blot, Neutralization
Classification Monoclonal
Clone 136318
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism
Host Species Mouse
Immunogen Pool of NS0-derived recombinant human FGF R3 alpha (IIIb) and Sf21-derived FGF R3 alpha (IIIc)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2261
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.