missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FGFR2 Split FISH Probe
Click to view available options
Quantity:
100 μL
200 μL
Dimensione della confezione:
100µL
200µL
Descrizione
Includes
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifica
Specifica
| Content And Storage | Store at 4°C in the dark. |
| Format | Tube |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections) |
| Includes | DAPI Counterstain (1500 ng/mL ) 250 uL |
| Product Type | FGFR2 Split FISH Probe |
| Quantity | 100 μL |
| Form | Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto