missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR2 Antibody (OTI5C5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70731PE
This item is not returnable.
View return policy
Description
FGFR2 Monoclonal specifically detects FGFR2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| FGFR2 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| FGFR2 | |
| Full length recombinant protein of human FGF R2 (NP_000132) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Flow Cytometry | |
| OTI5C5 | |
| Flow Cytometry | |
| BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis, Protein Kinase | |
| 2263.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction