missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FGFR2 Monoclonal antibody specifically detects FGFR2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | FGFR2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI5C5 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human FGFR2(NP_000132) produced in HEK293T cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?