missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR2 Antibody (98739), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB6843A
This item is not returnable.
View return policy
Description
FGFR2 Monoclonal antibody specifically detects FGFR2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF
Specifications
| FGFR2 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
| 98739 | |
| Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready | |
| BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF R2 isoforms and mouse myeloma cell line NS0-derived recombinant human FGF R2 isoforms | |
| 0.1 mL | |
| Angiogenesis, Protein Kinase | |
| 2263 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction