missing translation for 'onlineSavingsMsg'
Learn More

FGFR2 alpha Antibody (1057909) [PerCP], Novus Biologicals™

Product Code. 30734850 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734850 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734850 Supplier Novus Biologicals Supplier No. FAB11119C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGFR2 alpha Monoclonal antibody specifically detects FGFR2 alpha in Human samples. It is validated for Flow Cytometry,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFR2 alpha
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1057909
Conjugate PerCP
Formulation PBS
Gene Alias BBDS;BEK;BFR-1;CD332;CEK3;CFD1;ECT1;Fibroblast growth factor receptor;Fibroblast growth factor receptor 2;JWS;KGFR;K-SAM;Receptor protein-tyrosine kinase;TK14;TK25
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human FGFR2 alpha protein, Arg22-Glu377, Accession # P21802.1
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2263
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.