Learn More
FGFR1OP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531050
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7, mouse stomach. Predicted reactivity: Mouse (98%), Rat (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May be involved in wound healing pathway.Specifications
FGFR1OP2 | |
Polyclonal | |
Unconjugated | |
FGFR1OP2 | |
1500031J01Rik; FGFR1 oncogene partner 2; FGFR1 oncogene partner 2 homolog; FGFR1OP2; fibroblast growth factor receptor 1 oncogene partner 2; HSPC123; HSPC123-like; Wit3.0; Wit3.0_alpha; Wit3.0_beta; wound inducible transcript 3.0; wound inducible transcript 3.0 protein; wound-inducible transcript 3.0 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26127, 67529 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9CRA9, Q9NVK5 | |
FGFR1OP2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 200 of Human FGFR1OP2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |