Learn More
FGFR1OP Mouse anti-Human, Unconjugated, Clone: 1F1D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67207-1-IG-20UL
This item is not returnable.
View return policy
Description
The FGFR1OP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FGFR1OP. This antibody recognizes human antigen. The FGFR1OP antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| FGFR1OP | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, ELISA | |
| 1F1D8 | |
| Unconjugated | |
| 20 μL | |
| 11116 | |
| IgG1 |