missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FGFR1 Monoclonal antibody specifically detects FGFR1 in Human, Mouse samples. It is validated for Western Blot, Neutralization
Specifications
Specifications
| Antigen | FGFR1 |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Conjugate | APC |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?