missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR1 Antibody (M2F12), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-829MFV500
This item is not returnable.
View return policy
Description
FGFR1 Monoclonal antibody specifically detects FGFR1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| FGFR1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| M2F12 | |
| Flow Cytometry | |
| basic fibroblast growth factor receptor 1, BFGFR, bFGF-R-1, CD331, CD331 antigen, CEK, EC 2.7.10, EC 2.7.10.1, FGFBR, FGFR-1, fibroblast growth factor receptor 1, FLGH3, FLJ99988, FLT-2, FLT2H4, Fms-like tyrosine kinase 2, fms-related tyrosine kinase 2, H2, H5, HBGFR, heparin-binding growth factor receptor, hydroxyaryl-protein kinase, KAL 2, KAL2, N-SAM, OGD, Proto-oncogene c-Fgr, soluble FGFR1 variant 1, soluble FGFR1 variant 2 | |
| Recombinant human ectodomain of FGFr1a expressed in E. coli beginning with pro23; antigen contained NH2-terminal gly-ser-pro-gly-ile and COOH-terminal glu-phe sequences. | |
| 0.1 mL | |
| Angiogenesis, Apoptosis, Core ESC Like Genes, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers | |
| 2260 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction