missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFBP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24725-1-AP-150UL
This item is not returnable.
View return policy
Description
FGFBP3 is a heparin-binding protein which binds to FGF2, prevents binding of FGF2 to heparin and probably inhibits immobilization of FGF2 on extracellular matrix glycosaminoglycans, allowing its release and subsequent activation of FGFR signaling which leads to increased vascular permeability.Specifications
| FGFBP3 | |
| Polyclonal | |
| Unconjugated | |
| FGFBP3 | |
| C10orf13, FGF binding protein 3, FGF BP3, FGFBP 3, FGFBP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 143282, 72514 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q1HCM0, Q8TAT2 | |
| FGFBP3 | |
| FGFBP3 Fusion Protein Ag20573 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction