missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFBP3 Rabbit anti-Human, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89733-100UL
This item is not returnable.
View return policy
Description
FGFBP3 Monoclonal specifically detects FGFBP3 in Human samples. It is validated for ELISA.
Specifications
| FGFBP3 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 143282 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 2 | |
| ELISA 1:5000-1:10000 | |
| C10orf13, Chromosome 10 Open Reading Frame 13, FGF-Binding Protein 3, FGFBP3, FGF-BP3, FGFBP-3, Fibroblast Growth Factor-Binding Protein 3 | |
| Recombinant Human FGFBP3 Protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction