missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFBP3 Antibody (002), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89733PCP
This item is not returnable.
View return policy
Description
FGFBP3 Monoclonal antibody specifically detects FGFBP3 in Human samples. It is validated for ELISA
Specifications
| FGFBP3 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 143282 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| 002 | |
| ELISA | |
| C10orf13, Chromosome 10 Open Reading Frame 13, FGF-Binding Protein 3, FGFBP3, FGF-BP3, FGFBP-3, Fibroblast Growth Factor-Binding Protein 3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human FGFBP3 (Uniprot#: Q8TAT2; Met1-Gly258). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction