missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FGFBP2 Polyclonal Antibody

Product Code. 15756561 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15756561 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15756561 Supplier Invitrogen™ Supplier No. PA558686

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PVLRPSVCRE AGPQAHMQQV TSSLKGSPEP NQQPEAGTPS LRPKATVKLT EATQLGKDSM EELGKAKPTT R Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 30%.

This gene encodes a member of the fibroblast growth factor binding protein family. The encoded protein is a serum protein that is selectively secreted by cytotoxic lymphocytes and may be involved in cytotoxic lymphocyte-mediated immunity. An increase in the amount of gene product may be associated with atopic asthma and mild extrinsic asthma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFBP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FGFBP2
Gene Accession No. Q9BYJ0
Gene Alias 37 kDa killer-specific secretory protein; FGF-binding protein 2; FGFBP2; FGF-BP2; FGFBP-2; fibroblast growth factor binding protein 2; fibroblast growth factor-binding protein 2; HBp17-related protein; HBP17RP; HBp17-RP; killer-specific secretory protein of 37 kDa; KSP37; UNQ425/PRO1065
Gene Symbols FGFBP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FGFBP2. Recombinant protein control fragment (Product #RP-96652).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83888
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.