missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGFBP1 Monoclonal Antibody (KAA123_3F10), Invitrogen™
Mouse Monoclonal Antibody
£453.00 - £760.00
Specifications
| Antigen | FGFBP1 |
|---|---|
| Clone | KAA123_3F10 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30203272
|
Invitrogen™
MA556347 |
200 μL |
£760.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30204452
|
Invitrogen™
MA555967 |
50 μL |
£453.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: DTLGNTQIKQ KSRPGNKGKF VTKDQANCRW AATEQEEGIS LKVECTQLDH EFSCVFAGNP TSCLKLKDER VYWKQVARNL RSQKDICRYS KTAVKTRVCR KDFPESSLKL VSSTLFGNTK PRKEKTEMSP REHIKGKETT
FGFBP1 Monoclonal antibody specifically detects FGFBP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| FGFBP1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 17 kDa HBGF-binding protein; 17 kDa heparin-binding growth factor-binding protein; FGF-binding protein 1; FGFBP; FGF-BP; Fgfbp1; FGF-BP1; FGFBP-1; fibroblast growth factor binding protein 1; fibroblast growth factor-binding protein 1; growth factor binding protein-1; growth factor-binding protein 1; HBP17; heparin-binding growth factor binding protein | |
| Recombinant human FGFBP1 (immunogen range is 41-180). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| FGFBP1 |
| KAA123_3F10 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q14512 | |
| 9982 | |
| IgG2b | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title