missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20711-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to FGF8.
Plays an important role in the regulation of embryonic development, cell proliferation, cell differentiation and cell migration. Required for normal brain, eye, ear and limb development during embryogenesis. Required for normal development of the gonadotropin-releasing hormone (GnRH) neuronal system. Plays a role in neurite outgrowth in hippocampal cells.Specifications
| FGF8 | |
| Polyclonal | |
| Unconjugated | |
| FGF8 | |
| AIGF, Androgen induced growth factor, FGF 8, FGF8, Fibroblast growth factor 8, HBGF 8, KAL6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14179, 2253, 29349 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P37237, P55075 | |
| FGF8 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction