Learn More
PeproTech FGF5 Monoclonal Antibody, PeproTech®
Mouse Monoclonal Antibody
Brand: PeproTech 500-M40-500UG
Description
AA Sequence of recombinant protein: MAWAHGEKRL APKGQPGPAA TDRNPIGSSS RQSSSSAMSS SSASSSPAAS LGSQGSGLEQ SSFQWSPSGR RTGSLYCRVG IGFHLQIYPD GKVNGSHEAN MLSVLEIFAV SQGIVGIRGV FSNKFLAMSK KGKLHASAKF TDDCKFRERF QENSYNTYAS AIHRTEKTGR EWYVALNKRG KAKRGCSPRV KPQHISTHFL PRFKQSEQPE LSFTVTVPEK KNPPSPIKSK IPLSAPRKNT NSVKYRLKFR FG. Preparation: Produced in BALB/c mice immunized with highly pure Recombinant Human FGF-5. Anti-Human FGF-5-specific antibody was purified from cell culture by Protein G affinity chromatography.
Plays an important role in the regulation of cell proliferation and cell differentiation. Required for normal regulation of the hair growth cycle. Functions as an inhibitor of hair elongation by promoting progression from anagen, the growth phase of the hair follicle, into catagen the apoptosis-induced regression phase.
Specifications
| FGF5 | |
| Monoclonal | |
| PBS with no preservative | |
| P12034 | |
| FGF5 | |
| E.coli-derived Recombinant Human FGF-5. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| Unconjugated | |
| FGF5 | |
| angora; FGF; FGF5; FGF-5; Fibroblast growth factor; Fibroblast growth factor 5; fibroblast growth factor FGF-5; go; HBGF-5; Heparin-binding growth factor 5; Smag-82; TCMGLY | |
| Mouse | |
| Protein G | |
| RUO | |
| 2250 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.