Learn More
PeproTech FGF4 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
£177.00 - £2745.00
Specifications
| Antigen | FGF4 |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17884213
|
PeproTech
500-P158BT-25UG |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17893933
|
PeproTech
500-P158BT-50UG |
50 μg |
£228.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17854513
|
Invitrogen
500-P158BT-1MG |
2 x 500 μg | |||||||
|
30054419
|
PeproTech
500-P158BT-500UG |
500 μg |
£1466.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: GRGGAAAPTA PNGTLEAELE RRWESLVALS LARLPVAAQP KEAAVQSGAG DYLLGIKRLR RLYCNVGIGF HLQALPDGRI GGAHADTRDS LLELSPVERG VVSIFGVASR FFVAMSSKGK LYGSPFFTDE CTFKEILLPN NYNAYESYKY PGMFIALSKN GKTKKGNRVS PTMKVTHFLP RL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human FGF-4. Anti-Human FGF-4-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hFGF-4 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human FGF-4 (500-P158) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hFGF-4. Western Blot: To detect hFGF-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hFGF-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P158BT-1MG will be provided as 2 x 500 μg
Fibroblast growth factor 4 (FGF4) is a member of the fibroblast growth factor (FGF) family that possess broad mitogenic and cell survival activities and play key roles in growth and survival of stem cells during embryogenesis, tissue regeneration, and carcinogenesis. FGF4 was identified by its strong oncogenic transforming activity and is a potent angiogenic factor, expressed in several highly vascularized tumors and also in adult mouse testis, intestine, and brain. Studies on the mouse homolog suggests a function in bone morphogenesis and limb development through the sonic hedgehog (SHH) signaling pathway. Furthermore, FGF4 regulates neural progenitor cell proliferation and neuronal differentiation. Recent studies show a growth-promoting role for FGF4 in human embryonic stem cells and a putative feedback inhibition mechanism by a novel FGF4 splice isoform that may serve to promote differentiation at a later stages of development.Specifications
| FGF4 | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Human | |
| P08620 | |
| 2249 | |
| E.coli-derived Recombinant Human FGF-4. | |
| Antigen affinity chromatography | |
| Fgf4 |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| FGF; FGF4; Fgf-4; Fgfk; Fibroblast growth factor; fibroblast growth factor 4; fibroblast growth factor 4 splice isoform; HBGF-4; heparin secretory transforming protein 1; heparin secretory-transforming protein 1; heparin-binding growth factor 1; heparin-binding growth factor 4; HST; Hst1; hst-1; HSTF1; HSTF-1; human stomach cancer, transforming factor from FGF-related oncogene; kaposi sarcoma oncogene; Kfgf; K-FGF; K-fibroblast growth factor; KS3; oncogene HST; transforming protein KS3 | |
| Fgf4 | |
| Primary | |
| -20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.