Learn More
Invitrogen™ Human FGF4 (Cell Lysates) ELISA Kit
Description
The Human Fibroblast Growth Factor 4 Cell Lysates (Hu FGF4CL) ELISA quantitates Hu FGF4CL in human cell and tissue lysates. The assay will exclusively recognize both natural and recombinant Hu FGF4CL. Principle of the method The Human FGF4CL solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P08620 |
| Assay Range | 24.69 to 18,000 pg/mL |
| Assay Sensitivity | 50 pg/mL |
| Calibration Range | 50 pg/mL to 18,000 pg/mL |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
| Product Type | ELISA |
| Protein Family | Growth Factors & Receptors |
| Sample Type | Cell Lysates |
| Specificity | The antibody pair provided in this kit recognizes human FGF-4. |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.