missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16874-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to FGF-3. It does not bind other FGFs.
Fibroblast growth factors (FGFs) can stimulate the proliferation of mesenchymal, epithelial and neuroectodermal cells. FGF3 belongs to the heparin binding growth factors family. Members of the family include Acidic FGF (also known as aFGF or FGF 1), basic FGF (bFGF or FGF 2), FGF 3 (also known as Int2) and FGF 4 (also known as hst or Kaposi FGF). FGF 4 was first revealed to be an oncoprotein in Kaposi′s sarcoma cells. Other family members include FGF 5, FGF 6, FGF 7 (keratinocyte growth factor, KGF), FGF 8 (AIGF), FGF 9 (GAF) and FGF 10. FGF3 is activated by proviral insertion of mouse mammary tumor virus (MMTV). FGF3 is responsible for most of breast maligcies.Specifications
| FGF3 | |
| Polyclonal | |
| Unconjugated | |
| FGF3 | |
| FGF 3, FGF3, Fibroblast growth factor 3, HBGF 3, INT2, Proto oncogene Int 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2248 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11487 | |
| FGF3 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction