missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF23 Recombinant Rabbit Monoclonal Antibody (8J2D9)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | FGF23 |
|---|---|
| Clone | 8J2D9 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30282530
|
Invitrogen™
MA558159 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30283134
![]() |
Invitrogen™
MA558160 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: YPNASPLLGS SWGGLIHLYT ATARNSYHLQ IHKNGHVDGA PHQTIYSALM IRSEDAGFVV ITGVMSRRYL CMDFRGNIFG SHYFDPENCR FQHQTLENGY DVYHSPQYHF LVSLGRAKRA FLPGMNPPPY SQFLSRRNEI PLIHFNTPIP RRHTRSAEDD SERDPLNVLK PRARMTPAPA SCSQELPSAE DNSPMASDPL GVVRGGRVNT HAGGTGPEGC RPFAKFI.
The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The product of this gene inhibits renal tubular phosphate transport. This gene was identified by its mutations associated with autosomal dominant hypophosphatemic rickets, an inherited phosphate wasting disorder. Abnormally high level expression of this gene was found in oncogenic hypophosphatemic osteomalacia, a phenotypically similar disease caused by abnormal phosphate metabolism. Mutations in this gene have also been shown to cause familial tumoral calcinosis with hyperphosphatemia.Specifications
| FGF23 | |
| 1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q9GZV9 | |
| 8074 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-251 of human FGF23 (NP_065689.1). | |
| Antibody |
| 8J2D9 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| FGF23 | |
| ADHR; FGF; Fgf23; FGF-23; FGFN; Fibroblast growth factor; Fibroblast growth factor 23; Fibroblast growth factor 23 C-terminal peptide; Fibroblast growth factor 23 N-terminal peptide; HPDR2; HYPF; Phosphatonin; PHPTC; tumor-derived hypophosphatemia inducing factor; Tumor-derived hypophosphatemia-inducing factor; UNQ3027/PRO9828 | |
| FGF23 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title