missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FGF22 Monoclonal Antibody (435008)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561351

Product Code. 30534800

  • £490.00 / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The mouse homolog of this gene was found to be preferentially expressed in the inner root sheath of the hair follicle, which suggested a role in hair development. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FGF22
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q9HCT0
FGF22
E. coli-derived recombinant human FGF-22, Thr23-Ser170.
500 μg
Primary
Human
Antibody
IgG2a
Neutralization, Western Blot
435008
Unconjugated
FGF22
2210414E06Rik; FGF; Fgf22; FGF-22; Fibroblast growth factor; Fibroblast growth factor 22; UNQ2500/PRO5800
Mouse
Protein A/G
RUO
27006
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.