missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF20 Monoclonal Antibody (K24028_13C3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556188
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: LGGLEGLGQQ VGSHFLLPPA GERPPLLGER RSAAERSARG GPGAAQLAHL HGILRRRQLY CRTGFHLQIL PDGSVQGTRQ DHSLFGILEF ISVAVGLVSI RGVDSGLYLG MNDKGELYGS EKLTSECIFR EQFEENWYNT YSSNIYKHGD TGRRYFVALN KDGTPRDGAR SKRHQKFTHF LPRPVDPERV PELYKDLLMY
FGF-20 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-20 signals through FGFR 2c and 3c, and is expressed during limb and brain development.
Specifications
| FGF20 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9NP95 | |
| FGF20 | |
| Recombinant human FGF20 (immunogen range is 11-210). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| K24028_13C3 | |
| Unconjugated | |
| FGF20 | |
| FGF; Fgf20; FGF-20; Fibroblast growth factor; Fibroblast growth factor 20; RHDA2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 26281 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction