missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF18 Antibody (CL7628) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88869MFV500
This item is not returnable.
View return policy
Description
FGF18 Monoclonal antibody specifically detects FGF18 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| FGF18 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| FGF-18ZFGF5, fibroblast growth factor 18, zFGF5 | |
| This antibody was developed using a synthetic peptide derived from O76093, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication | |
| 8817 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL7628 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction