Learn More
PeproTech FGF17 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
Brand: PeproTech 500-P152GBT-50UG
Description
AA Sequence of recombinant protein: MTQGENHPSP NFNQYVRDQG AMTDQLSRRQ IREYQLYSRT SGKHVQVTGR RISATAEDGN KFAKLIVETD TFGSRVRIKG AESEKYICMN KRGKLIGKPS GKSKDCVFTE IVLENNYTAF QNARHEGWFM AFTRQGRPRQ ASRSRQNQRE AHFIKRLYQG QLPFPNHAEK QKQFEFVGSA PTRRTKRTRR PQPLT. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human FGF-17. Anti-Human FGF-17-specific antibody was purified by affinity chromatography and then biotinylated. Direct ELISA: To detect Human FGF-17 by direct ELISA (using 100 μL/well) a concentration of approximately 1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody allows the detection of at least 2000-4000 pg/mL of Recombinant Human FGF-17. Western Blot: To detect hFGF-17 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hFGF-17 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P152GBT-1 mg will be provided as 2 x 500 μg
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumor growth and invasion. This gene was shown to be prominently expressed in the cerebellum and cortex. The mouse homolog of this gene was localized to specific sites in the midline structures of the forebrain, the midbrain-hindbrain junction, developing skeleton and developing arteries, which suggests a role in central nervous system, bone and vascular development. This gene was referred to as FGF-13 in reference 2, however, its amino acid sequence and chromosomal localization are identical to FGF17.
Specifications
| FGF17 | |
| Polyclonal | |
| Biotin | |
| FGF17 | |
| FGF; FGF-13; FGF17; FGF-17; Fibroblast growth factor; Fibroblast growth factor 17; HH20; UNQ161/PRO187 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 8822 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| O60258 | |
| FGF17 | |
| E.coli-derived Recombinant Human FGF-17. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.