missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF16 Monoclonal Antibody (K52035_6E10), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | FGF16 |
|---|---|
| Clone | K52035_6E10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30204174
|
Invitrogen™
MA556137 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30211804
|
Invitrogen™
MA555757 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: LDWDLHGFSS SLGNVPLADS PGFLNERLGQ IEGKLQRGSP TDFAHLKGIL RRRQLYCRTG FHLEIFPNGT VHGTRHDHSR FGILEFISLA VGLISIRGVD SGLYLGMNER GELYGSKKLT RECVFREQFE ENWYNTYAST LYKHSDSERQ YYVALNKDGS PREGYRTKRH QKFTHFLPRP VDPSKLPSMS
FGF16 Monoclonal antibody specifically detects FGF16 in Human samples. It is validated for Western BlotSpecifications
| FGF16 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| FGF; FGF16; FGF-16; Fibroblast growth factor; Fibroblast growth factor 16; metacarpal 4-5 fusion; MF4 | |
| Recombinant human FGF16 (immunogen range is 11-200). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| FGF16 |
| K52035_6E10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O43320 | |
| 8823 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title