missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101999
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FGF13.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. This gene is located in a region on chromosome X, which is associated with Borjeson-Forssman-Lehmann syndrome (BFLS), making it a possible candidate gene for familial cases of the BFLS, and for other syndromal and nonspecific forms of X-linked mental retardation mapping to this region. Alternative splicing of this gene at the 5' end results in several transcript variants encoding different isoforms with different N-termini.
Specifications
| FGF13 | |
| Polyclonal | |
| Unconjugated | |
| FGF13 | |
| AFGF; ECGF; ECGFA; ECGFB; FGF; FGF 1; FGF alpha; FGF13; FGF-13; FGF13A; FGF2; FGFA; Fhf2; FHF-2; Fibroblast growth factor; fibroblast growth factor 13; Fibroblast growth factor homologous factor 2; GLIO703; HBGF 1; HBGF1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14168, 2258, 84488 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70377, Q92913, Q9ERW3 | |
| FGF13 | |
| A synthesized peptide derived from human FGF13(Accession Q92913), corresponding to amino acid residues V153-M203. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction