missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FGF12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100815
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FGF12.
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. This growth factor lacks the N-terminal signal sequence present in most of the FGF family members, but it contains clusters of basic residues that have been demonstrated to act as a nuclear localization signal. When transfected into mammalian cells, this protein accumulated in the nucleus, but was not secreted. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| FGF12 | |
| Polyclonal | |
| Unconjugated | |
| Fgf12 | |
| AV114868; B230343J05Rik; FGF; FGF12; FGF-12; FGF12B; FHF1; FHF-1; Fhf1b; Fibroblast growth factor; Fibroblast growth factor 12; fibroblast growth factor 12B; fibroblast growth factor FGF-12b; fibroblast growth factor homologous factor 1; fibroblast growth factor homologous factor 1b; myocyte-activating factor | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14167, 170630, 2257 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P61150, P61328, P61329 | |
| Fgf12 | |
| A synthesized peptide derived from human FGF12(Accession P61328), corresponding to amino acid residues K201-T243. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction