missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FGF1 (FGF alpha) ELISA Kit

Product Code. 17155439
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Unit Size:
1 set
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 17155439

Brand: Invitrogen™ EHFGF1X5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Human Fibroblast Growth Factor 1 (Hu FGF1/FGFα) ELISA quantitates Hu FGF1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FGF1. Principle of the method The Human FGF1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

FGF1 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05230
Assay Range 16.38 to 4000 pg/mL
Assay Sensitivity 12 pg/mL
Calibration Range 12 pg/mL to 4000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family Growth Factors & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human BLC, BDNF, bFGF, BMP-4, BMP-5, BMP-7, b-NGF, Eotaxin-1, Eotaxin-2, FGF-4, FGF-7, G-CSF, GM-CSF, I-309, ICAM-1, IFN-gamma, IL-1 alpha, IL-1 beta, IL-1 ra, IL-2, IL-4, IL-5, IL-6, IL-6 sR, IL-7, IL-8, IL-10, IL-11, IL-12 p40, IL-12 p70, IL-13, IL-15, IL-16, IL-17, MCP-1, M-CSF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PDGF-BB, RANTES, sTNFRI, TIMP-1, TIMP-2, TNF alpha, TNF beta, VEGF, VEGFR2, VEGFR3.
Sufficient For 5 x 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2246
Gene Symbol AFGF,ECGF,ECGF-beta,ECGFA,ECGFB,FGF-1,FGF-alpha,FGFA,GLIO703,HBGF-1,HBGF1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Angiogenesis
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym AFGF, ECGF, ECGF-beta, ECGFA, ECGFB, FGF-1, FGF-alpha, FGFA, GLIO703, HBGF-1, HBGF1
Target FGF1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.