missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF basic/FGF2/bFGF Antibody (FB-8), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1128AF750
This item is not returnable.
View return policy
Description
FGF basic/FGF2/bFGF Monoclonal specifically detects FGF basic/FGF2/bFGF in Human, Bovine samples. It is validated for Western Blot, ELISA.
Specifications
| FGF basic/FGF2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| FGF2 | |
| Recombinant full length protein (Human). | |
| Primary | |
| Does not cross-react with human fibroblast growth factor - acidic (aFGF). | |
| Store at 4°C in the dark. |
| Western Blot, ELISA | |
| FB-8 | |
| Western Blot, ELISA | |
| Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 | |
| Mouse | |
| 0.1 mL | |
| 2247 | |
| Human, Bovine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction