missing translation for 'onlineSavingsMsg'
Learn More

FGF acidic/FGF1 Antibody (OTI4D2), DyLight 594, Novus Biologicals™

Product Code. 18408412 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408412 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408412 Supplier Novus Biologicals Supplier No. NBP270717DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGF acidic/FGF1 Monoclonal antibody specifically detects FGF acidic/FGF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF acidic/FGF1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone OTI4D2
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias AFGF, alpha, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 16-155 of human FGF acidic/FGF1 (NP_000791) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2246
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.