missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-8 Antibody (001), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90327CL1
This item is not returnable.
View return policy
Description
FGF-8 Monoclonal antibody specifically detects FGF-8 in Human samples. It is validated for ELISA
Specifications
| FGF-8 | |
| Monoclonal | |
| CoraFluor 1 | |
| AIGFKAL6, Androgen-induced growth factor, FGF-8, fibroblast growth factor 8, fibroblast growth factor 8 (androgen-induced), HBGF-8, Heparin-binding growth factor 8, MGC149376 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human FGF-8 (Accession#: NP_149355.1; Gln23-Arg204). | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication | |
| 2253 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 001 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction