missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-3 Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | FGF-3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18307984
|
Novus Biologicals
NBP3-15491-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18358115
|
Novus Biologicals
NBP3-15491-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGF-3 Polyclonal antibody specifically detects FGF-3 in Rat samples. It is validated for Western BlotSpecifications
| FGF-3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| fibroblast growth factor 3, fibroblast growth factor 3 (murine mammary tumor virus integration site (v-int-2) oncogene homolog), HBGF-3, mouse, oncogene INT2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FGF-3 (NP_005238.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Tumor Suppressors | |
| PBS, 50% glycerol, pH7.3 | |
| 2248 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title