missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-3 Antibody (MSD1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB200-603AF488
This item is not returnable.
View return policy
Description
FGF-3 Monoclonal specifically detects FGF-3 in Human, Mouse, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.
Specifications
| FGF-3 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FGF3 | |
| Peptide conjugated to BSA. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Tumor Suppressors | |
| 2248 | |
| Human, Mouse, Xenopus species | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MSD1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| fibroblast growth factor 3, fibroblast growth factor 3 (murine mammary tumor virus integration site (v-int-2) oncogene homolog), HBGF-3, mouse, oncogene INT2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody recognizes all naturally occurring FGF-3 variants and all recombinant FGF-3 proteins. | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction