missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FGF-23 Monoclonal specifically detects FGF-23 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
Specifications
| Antigen | FGF-23 |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
| Classification | Monoclonal |
| Clone | FGF23/4166 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?