missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-23 Mouse anti-Human, Clone: FGF23/4166, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07371-100ug
This item is not returnable.
View return policy
Description
FGF-23 Monoclonal specifically detects FGF-23 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
| FGF-23 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor | |
| Recombinant fragment (around aa25-251) of human FGF-23 protein (exact sequence is proprietary) | |
| 100 μg | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 8074 | |
| Store at 4°C. | |
| IgG2a κ |
| ELISA, Peptide Array | |
| FGF23/4166 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction