missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-23 Mouse anti-Human, Clone: FGF23/4163, Novus Biologicals™
Mouse Monoclonal Antibody
£221.00
Specifications
| Antigen | FGF-23 |
|---|---|
| Clone | FGF23/4163 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
FGF-23 Monoclonal specifically detects FGF-23 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| FGF-23 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 8074 | |
| IgG1 κ | |
| Protein A or G purified |
| FGF23/4163 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor | |
| Recombinant fragment (around aa25-251) of human FGF-23 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title