missing translation for 'onlineSavingsMsg'
Learn More

FGF-23 Antibody (FGF23/4168) - Azide and BSA Free, Novus Biologicals™

Product Code. 18352162 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352162 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18352162 Supplier Novus Biologicals Supplier No. NBP308383

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FGF-23 Monoclonal specifically detects FGF-23 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF-23
Applications Immunohistochemistry (Paraffin), Peptide Array
Classification Monoclonal
Clone FGF23/4168
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 ug/mL, Protein Array
Formulation 10mM PBS
Gene Alias ADHR, FGF-23, fibroblast growth factor 23, HPDR2, HYPF, Phosphatonin, PHPTC, tumor-derived hypophosphatemia inducing factor, Tumor-derived hypophosphatemia-inducing factor
Host Species Mouse
Immunogen Recombinant fragment (around aa25-251) of human FGF-23 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8074
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.