Learn More
FGF-21 Antibody (CL6491) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76496APC
Product Code. 30289398
  • £558.00 / 0.10mL
Description
FGF-21 Monoclonal antibody specifically detects FGF-21 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGF-21
Monoclonal
APC
FGF-21, fibroblast growth factor 21
This antibody was developed using a synthetic peptide derived from Q9NSA1, with the exact immunogen sequence remaining proprietary.
0.1 mL
Angiogenesis, Biologically Active Proteins, Cancer
26291
Store at 4⪚C in the dark.
IgG1
Western Blot
CL6491
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified