missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-21 Antibody (461804) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB25372JF585
This item is not returnable.
View return policy
Description
FGF-21 Monoclonal antibody specifically detects FGF-21 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| FGF-21 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| FGF-21, fibroblast growth factor 21 | |
| E. coli-derived recombinant human FGF-21, His29-Ser209, Accession # Q9NSA1 | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer | |
| 26291 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 461804 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction